missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ViewRNA™ Tissue Positive Control Kit (2-plex) (4 rat kidney slides + probes)

Product Code. 31722876
Click to view available options
Quantity:
1 Kit
Unit Size:
Each

Product Code. 31722876

Brand: Invitrogen™ QVT0516

Please to purchase this item. Need a web account? Register with us today!

The ViewRNA Tissue Positive Control Kit (2-Plex) is designed for use with the ViewRNA Tissue Assay Core Kit, 1-plex or ViewRNA Tissue Assay Blue Module.

The ViewRNA Tissue Positive Control Kit (2-Plex) is designed for use with the ViewRNA Tissue Assay Core Kit, 1-plex (Cat. No. 19931) or ViewRNA Tissue Assay Blue Module (19932). This positive control kit includes 4-6 μm thick rat kidney FFPE sections attached to positively charged slides as well one vial each of TYPE 1 rat Spp1 and TYPE 6 rat Ubc probe sets.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 4 to 6 μm thick rat kidney FFPE sections attached to positively charged slides, 4 slides
• TYPE 1 rat Spp1 probe set, 1 vial
• TYPE 6 rat Ubc probe set, 1 vial
Format Kit
For Use With (Application) Tissue Analysis
For Use With (Equipment) ViewRNA Tissue Assay Core Kit, 1-plex (Cat. No. 19931) or ViewRNA Tissue Assay Blue Module (19932)
Product Type RNA In Situ Hybridization Tissue Positive Control Kit
Quantity 1 Kit

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.