Learn More
Gilson™ Position Rack Flask Vessel Type
The racks come in a variety of configurations to accommodate many different vials, tubes, microtubes, and multi-well microplates
Brand:  Gilson™ 152422
Additional Details : Weight : 0.45000kg
 View more versions of this product
Product Code. 16626125
  • £407.04 / Pack of 5
Estimated Shipment: 29-07-2024
to see stock.
Add to basket
Description
Description
  • These various racks are used with current and legacy Gilson liquid handlers, ASPEC™ systems, and other instruments
  • Flask Vessel Type
Specifications
Specifications
Code 21
9 mL Vial (13 x 100 mm)
5 Pk.
Flask
60