Learn More
Smiths Medical Portex Tubing Adaptor
Product Code. 13170873
Encompass
Product Code. 13170873
Brand: Smiths Medical 700/180/700
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Luer female to luer male connector, 700/180, NS standard taper for male and female ends ensures smooth catheter connections, 10 per box male luer OD4.4mm; female luer ID4.3mm max Smiths Medical