missing translation for 'onlineSavingsMsg'
Learn More

Portex™Polypropylene Y-shape Tubing Connector

Product Code. 12780038
Encompass
Change view
Click to view available options
Diameter (Metric) Inner:
7.5 mm
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner unitSize
12780038 7.5 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12780038 Supplier Portex Supplier No. 700/140/005

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Order Info

DiameterMetricInner

Specifications

Diameter (Metric) Inner 7.5 mm
Material Polypropylene
For Use With For 7.5 I.D. x 11.5mm O.D. Tubing
Packaging Quantity 10 Pack
Shape Y
Product Type Tubing Connector
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.