missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ POROS™ XS Strong Cation Exchange Resin

POROS XS is the first high-capacity, high-resolution resin that allows loading to >100 mg/mL in the presence of 15 mS/cm NaCl, while delivering unprecedented separation capability

£167.00 - £564.00

Specifications

Content And Storage Store in refrigerator (2–8°C).
Format Poros Bead
Product Line POROS™
Purification Target Protein
Shipping Condition Room Temperature
View More Specs

Alert:

For Research Use Only. Not for use in diagnostic procedures.


Products 2
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
15622135
View Documents Promotion Details
Thermo Scientific™
82071
10 mL
£167.00
10mL
View alternative products
Save Up to 
»
Estimated Shipment: 17-05-2024
to see stock.
Add to basket
 
15632135
View Documents Promotion Details
Thermo Scientific™
82072
50 mL
£564.00
50mL
View alternative products
Save Up to 
»
Estimated Shipment: 13-05-2024
to see stock.
Add to basket
 
Description

Description

POROS™ XS Strong Cation Exchange Resin is the first high-capacity, high-resolution resin that allows loading to more than 100 mg/mL capacity in the presence of up to 15 mS/cm NaCl, while delivering unprecedented separation capability. A major improvement on traditional cation exchangers, POROS XS resin sets the standard for cation exchange chromatography.

  • Dynamic binding capacity leads to reduced column size, smaller footprint, decreased buffer usage, and reduced cycling
  • High resolution supports better product yield and improved impurity clearance
  • Robust salt tolerance allows improved process flexibility, alignment, and process flow streamlining
  • Low back pressure and linear pressure responses help improve scalability
  • Rigid polymeric bead with covalent surface chemistry makes for easier handling and packing, and superior physical and chemical stability, resulting in a robust manufacturing process

Dynamic binding capacityPOROS XS resin has a high binding capacity of more than 100 mg/mL, at 5% breakthrough, over a broad range of process conditions.Higher dynamic binding capacity was achieved by optimizing the base bead pore structure and surface chemistry.

High resolution

Using an optimized 50 µm particle size, POROS XS resin maintains the superior resolving capacity of POROS™ HS resin while overcoming the challenging impurity levels associated with high titer processes. Resolution is consistent across a wide range of operating flow rates and is maintained independent of protein load.

Robust salt tolerance
POROS XS resin has high and consistent protein capacity across a broad range of salt concentrations up to 150 mM NaCl (15 mS/cm conductivity). The ability to load columns under higher conductivity reduces or helps eliminate the dilution of column feed streams or the removal of unit operations, such as tangential flow filtration. The optimized surface ionic capacity of POROS XS resin minimizes competitive binding between the protein of interest and the counterion, resulting in superior salt tolerance.

Specifications

Specifications

Store in refrigerator (2–8°C).
POROS™
Room Temperature
XS Strong Cation Exchange Resin
Cation Exchange Chromatography
Poros Bead
Protein
XS
Crosslinked Poly(styrene-divinylbenzene)
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.