missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ POROS™ CaptureSelect™ IgA Affinty HPLC Columns

Product Code. 30019350 Shop All Thermo Scientific Products
Click to view available options
Dimensions:
4.6 x 100 mm
4.6 x 50 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 30019350

Brand: Thermo Scientific™ 4485170

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ POROS™ CaptureSelect™ IgA HPLC columns incorporate the advantages of high-speed POROS chromatography resins with the unique selectivity of CaptureSelect ligands.

Analytical groups supporting biologic drug development require rapid, sensitive, and precise methods for protein quantitation from cell culture, fermentations broths, or purification intermediates. It is critical for biopharmaceutical manufacturers to be able to monitor bioreactor product titers to determine when to harvest or abort a reactor cycle. POROS CaptureSelect IgA HPLC columns incorporate the advantages of high-speed POROS chromatography resins with the unique selectivity of CaptureSelect ligands. The combination allows three-minute quantitation of IgA.

  • Broad utility—POROS A, G, and CaptureSelect affinity ligands address most biological drug classes
  • Rapid—POROS resins offer fast quantitation or purification of immunoglobulins, fusion proteins
  • Hands-off operation—HPLC method can be automated for streamlined operation
  • Robust—allows for high-performance purification over hundreds of chromatography cycles

High Speed with High Specificity

The combination of CaptureSelect IgA affinity ligands and POROS 20 μm resin provides a high flow rate, high performance chromatography solution for the rapid titer determinations and small-scale purification of IgA.

POROS high performance chromatography allows for the quantitation of product from clarified harvest or partially purified intermediates—typically in under three minutes. Moreover, only minimal sample preparation (0.2 μm filtration) is required, and allows for an eluted product that is highly purified and suitable for further analysis, including mass spectrometry.

Automation

POROS affinity chromatography columns are suitable for use with HPLC systems, which allows for the automation of chromatography methods, data collection, and data analysis. In addition, the immobilized affinity ligands enable one method and buffer system to be used across a class of molecules, streamlining analytical approaches.

Multiple Cycles

Proper sample preparation, adherence to column operating instructions, and the use of an optimized column cleaning regime can result in strong re-use performance. POROS affinity chromatography columns can be cycled hundreds of times for cost-effective and reproducible quantitation or purification.

Specifications

Diameter (Metric) 4.6 mm
Length (Metric) 100 mm
Max. Pressure 2600 psi, 180 bar
Particle Size 20 μm
Pore Size 2000 Å
Quantity 1 Column
Stationary Phase IgA affinity ligand, human-specific
Content And Storage Column includes EZ grip fittings and column test certificate.
Store at 2°C to 8°C. DO NOT FREEZE.
Product Line CaptureSelect, POROS
Binding Capacity 7.6 mg/mL
Elution Volume Max 29 column volumes
Elution Volume Min 14 column volumes
High-throughput Compatibility Automated Protocols
Target Molecule Protein
Matrix POROS
Dimensions 4.6 x 100 mm
Column Type Affinity
Material PEEK
Type Column
Purification Target Protein
For Use With (Application) FPLC
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.