Learn More
Thermo Scientific™ POROS™ CaptureSelect™ HSA Affinity Column, 2.1 x 30 mm
Shop All Thermo Scientific ProductsBeskrivelse
Analytical groups supporting biologic drug development require rapid, sensitive and precise methods for protein quantitation from cell culture, fermentation harvests, or purification intermediates. It is critical for biopharmaceutical manufacturers to be able to monitor bioreactor product titers to determine when to harvest or abort a reactor cycle.
POROS CaptureSelect HSA HPLC columns incorporate the advantages of high-speed Poros chromatography resins with the unique selectivity of CaptureSelect ligands. The combination allows three minute quantitation of HSA fusion proteins.
Features:
- Broad utility—POROS A, G and CaptureSelectaffinity ligands address most biological drug classes
- Rapid—POROS resins offer fast quantitation or purification of immunoglobulins and fusion proteins
- Hands-off operation—HPLC method can be automated for streamlined operation
- Robust—deliver high-performance purification over hundreds of chromatography cycles
High Speed with High Specificity
The combination of CaptureSelect HSA affinity ligands when combined with POROS 20 μm resin provides a high flow rate, high performance chromatography solution for the rapid quantitation and small-scale purification of HSA Fusion proteins.
POROS high performance chromatography allows for the quantitation of product from clarified harvest or partially purified intermediates—typically in under three minutes. Moreover, only minimal sample preparation (0.2 μm filtration) is required, and the eluted product is highly purified and suitable for further analysis including mass spectrometry.
Automation
POROS affinity chromatography columns are suitable for use with HPLC systems, which allows the automation of chromatography methods, data collection, and data analysis. In addition, the immobilized affinity ligands enable one method and buffer system to be used across a class of molecules, streamlining analytical approaches.
Multiple Cycles
Proper sample preparation, adherence to column operating instructions, and the use of an optimized column cleaning regime results in strong re-use performance. POROS Affinity Chromatography columns can be cycled hundreds of times for cost-effective and reproducible quantitation or purification.
HSA Column Specifications:
- Protein Binding Capacity: 2.6 mg/mL
- Species Specificity: Human
- Column Material: PEEK™ polymer or stainless steel
- Frit: 2 μm, titanium
- POROS Resin: Cross-linked poly (styrenedivinylbenzene)
- CaptureSelect Ligands: Single-domain monospecific antibody fragment produced in Saccharomyces cerevisiae
Tekniske data
Tekniske data
| Diameter (Metric) | 2.1 mm |
| Flow Rate | Up to 5000 cm/hr. |
| Length (Metric) | 30 mm |
| Max. Pressure | 180 bar |
| Particle Size | 20 μm |
| Pore Size | 0.16 micron |
| Quantity | 1 Column |
| Stationary Phase | CaptureSelect™ HSA |
| Content And Storage | Column includes EZ grip fittings and column test certificate Store at 4°C. DO NOT FREEZE. |
| Product Line | CaptureSelect, POROS |
| Vis mere |
For Research Use Only. Not for use in diagnostics procedures.
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.