missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ POROS™ CaptureSelect™ FSH Affinity HPLC Columns

Product Code. 18496558 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
30 mm
50 mm
100 mm
Diameter (Metric):
2.1 mm
4.6 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Unit Size
18496558 50 mm 4.6 mm Each
18416428 30 mm 2.1 mm Each
18466648 100 mm 4.6 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18496558 Supplier Thermo Scientific™ Supplier No. 4481824
  • ONLINE EXCLUSIVE PRICE
    £2625.00
    £2455.00 / Pack of 1
    Save £170.00
    6% Off
View all promotions
 Restricted Item
Estimated Shipment: 17-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Analytical teams that support biologic drug development require rapid, sensitive, and precise methods for protein quantitation from cell culture, fermentations broths, or purification intermediates.

Analytical teams that support biologic drug development require rapid, sensitive, and precise methods for protein quantitation from cell culture, fermentations broths, or purification intermediates. It is critical for biopharmaceutical manufacturers to be able to monitor bioreactor product titers to determine when to harvest or abort a reactor cycle. POROS CaptureSelect FSH HPLC columns incorporate the advantages of high-speed POROS chromatography resins with the unique selectivity of CaptureSelect ligands, enabling 3-minute quantitation of human follicle stimulating hormone (FSH).

Features of this class of chromotography columns:

  • Broad utility—POROS A, G, and CaptureSelect affinity ligands address most biological drug classes
  • Rapid—POROS resins offer fast quantitation or purification of immunoglobulins and fusion proteins
  • Hands-off operation—HPLC method can be automated for streamlined operation
  • Robust—delivers high-performance purification over hundreds of chromatography cycles

High Speed with High Specificity

The combination of CaptureSelect FSH affinity ligand with POROS 20 μm resin provides a high flow-rate, high performance chromatography solution for the rapid quantitation and small-scale purification of FSH.

POROS high performance chromatography allows for the quantitation of product from clarified harvest or partially purified intermediates—typically in less than 3 minutes. Moreover, only minimal sample preparation (0.2 μm filtration) is required, and the eluted product is highly purified and suitable for further analysis, including mass spectrometry.

Automation

POROS affinity chromatography columns are suitable for use with HPLC systems, allowing for the automation of chromatography, data collection, and data analysis. In addition, the immobilized affinity ligands enable one method and buffer system to be used across a class of molecules, streamlining analytical approaches.

Multiple Cycles

Proper sample preparation, adherence to column operating instructions, and the use of an optimized column cleaning regime results in strong re-use performance. POROS affinity chromatography columns can be cycled hundreds of times for cost-effective and reproducible quantitation or purification.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 4.6 mm
Includes Column includes EZ grip fittings and column test certificate.
Length (Metric) 50 mm
Max. Pressure 180 bar
Particle Size 20 μm
Pore Size 2000 Å
Quantity 1 Column
Stationary Phase Follicle Stimulating Hormone affinity ligand, humanspecific
Product Line CaptureSelect, POROS
Binding Capacity 1.5 mg/mL resin
Elution Volume Max <800 μL
High-throughput Compatibility Automated Protocols
Sample Type Biopharmaceutical Purification Sample, Bulk Drug Substance Sample, Cell Cultures
Target Molecule Follicle Stimulating Hormone
Volume (Metric) 0.8 mL
Column Type Affinity
Phase Affinity Separation
Material PEEK
Purification Target Follicle Stimulating Hormone
Technique Antibody Affinity Chromatography
For Use With (Application) FPLC
Product Type Column
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.