missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ POROS™ AAVX RoboColumn™ Pre-packed columns

Product Code. 18406538 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
200 μL
600 μL
Unit Size:
200µL
600µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
18406538 200 μL 200µL
18406448 600 μL 600µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18406538 Supplier Thermo Scientific™ Supplier No. A37794

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ RoboColumn™ chromatography products are small columns provided in 8-column strips.

RoboColumn™ chromatography products are small columns provided in 8-column strips. They are useful for fully automated and parallel chromatographic separations using robotic liquid handling platforms.

These RoboColumn™ columns are pre-packed with POROS CaptureSelect AAVX Affinity Resin (Cat. No. A36743), engineered to address the high selectivity and capacity requirements for downstream purification of a broad range of naturally occurring and synthetic adeno-associated virus (AAV) serotypes used for gene therapy applications. POROS CaptureSelect AAVX resin has demonstrated binding reactivity towards a set of AAV serotypes that includes AAV1 to AAV8 and AAVrh10, as well as synthetic serotypes.

POROS CaptureSelect AAVX resin enables a robust, fast, and platformable purification process for wide-ranging AAV serotypes with excellent purity and yield obtained in one step.

Features of POROS CaptureSelect AAVX resin include:

  • One-step AAV purification from crude material with high purity and yield
  • High specificity and capacity, maximizing yield by significantly reducing process steps
  • Superior capacity maintained at high flow rates
  • Excellent scalability and easier handling and packing due to rigid polymeric bead
  • Non-animal derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

A wide selection of RoboColumn™ is available, including columns pre-packed with POROS ion exchange, HIC, and CaptureSelect affinity resins. All POROS high performance chromatography resins have a unique pore structure that provides excellent mass transport for high capacity and resolution over a wide range of flow rates.

RoboColumn™ is a registered trademark of Repligen Corporation.

Specifications

For Use With Robotic Liquid Handling
Particle Size 50 μm
Pore Size 100 nm
Quantity 200 μL
Stationary Phase Immobilized CaptureSelect™ AAVX Ligand
Content And Storage Shipped in 18% ethanol. Store at 2°C to 8°C.
Product Line POROS CaptureSelect
Volume (Metric) 200 μL
Column Type RoboColumn
Purification Target AAV
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.