missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PorM VHH-8His-Cys-tag Recombinant Alpaca Monoclonal Antibody (SAA0959)
Description
Compatible with Direct ELISA.
PorM is a component of the Type IX Secretion System (T9SS) in bacteria, facilitating the translocation of proteins across the outer membrane.
Specifications
Specifications
| Antigen | PorM VHH-8His-Cys-tag |
| Applications | ELISA |
| Classification | Recombinant Monoclonal |
| Clone | SAA0959 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene Alias | PGN_1674; porM |
| Host Species | Alpaca |
| Immunogen | Recombinant Porphyromonas gingivalis porM. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?