missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Porimin Antibody (CL14381), Novus Biologicals™
Mouse Monoclonal Antibody
£336.00 - £483.00
Specifications
| Antigen | Porimin |
|---|---|
| Clone | CL14381 |
| Concentration | 1 mg/mL |
| Dilution | Immunohistochemistry 1:20 - 1:50 |
| Applications | Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043430
|
Novus Biologicals™
NBP3-26852-25ul |
25 μL |
£336.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043807
|
Novus Biologicals™
NBP3-26852-100ul |
100 μL |
£483.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Porimin Monoclonal antibody specifically detects Porimin in Human samples. It is validated for ImmunohistochemistrySpecifications
| Porimin | |
| 1 mg/mL | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Apoptosis | |
| PBS, pH 7.2, 40% glycerol | |
| 114908 | |
| IgG2b | |
| Protein A purified |
| CL14381 | |
| Immunohistochemistry 1:20 - 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| KCT-3, KCT3keratinocytes associated transmembrane protein 3, Keratinocytes-associated transmembrane protein 3, PORIMINporimin, pro oncosis receptor inducing membrane injury, Pro-oncosis receptor inducing membrane injury, serine/threonine-rich receptor, transmembrane protein 123PORMIN | |
| This antibody was developed against a Synthetic Peptide of Q8N131. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title