missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems Porcine Luminex™ Discovery Assay

Product Code. 18742394
Change view
Click to view available options
Quantity:
1 Plex
10 Plex
11 Plex
12 Plex
13 Plex
14 Plex
15 Plex
16 Plex
17 Plex
18 Plex
19 Plex
2 Plex
20 Plex
3 Plex
4 Plex
5 Plex
6 Plex
7 Plex
8 Plex
9 Plex
Unit Size:
1 set
Product Code. Quantity unitSize
18742394 15 Plex 1 set
18751954 14 Plex 1 set
18732024 9 Plex 1 set
18732464 4 Plex 1 set
18742424 5 Plex 1 set
18732354 17 Plex 1 set
18762284 13 Plex 1 set
18792074 6 Plex 1 set
18722354 16 Plex 1 set
18732164 10 Plex 1 set
18722154 12 Plex 1 set
18752054 18 Plex 1 set
18722304 3 Plex 1 set
18762224 19 Plex 1 set
18742054 8 Plex 1 set
18741934 20 Plex 1 set
18742164 11 Plex 1 set
18762354 7 Plex 1 set
18782324 1 Plex 1 set
18772404 2 Plex 1 set
20 options
This item is not returnable. View return policy

Product Code. 18742394

Brand: R&D Systems LXSAPM15

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 15 Plex
Species Porcine
Binding Target Multiplex Cytokine Assay Kits
Content And Storage Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.