missing translation for 'onlineSavingsMsg'
Learn More

Porcine Liver Esterase, TRC

Product Code. 30383294
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383294 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30383294 Supplier Toronto Research Chemicals Supplier No. TRCE337455100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Porcine Liver Esterase
CAS 9016-18-6
Quantity 100 mg
Synonym Carboxylesterase, 1,4-Butanediol Diacrylate Esterase, 7-Amino-3-methoxy-3-cephem-4-carboxyl Ester Hydrolase, Aliesterase, Aminoacyl Esterase, B-Esterase, Biocatalytics 123, Butyrate Esterase, Butyryl Esterase, C4 Esterase, Carbonic Esterase, Carboxyesterase, Carboxyl Ester Hydrolase, Carboxyl Ester Lipase, Carboxyl Esterase, Carboxylate Esterase, Carboxylesterase 1, Carboxylesterase 2, Carboxylesterase B, Carboxylesterase ES-1, Carboxylesterases, Carboxylic Acid Esterase, Carboxylic Ester Hydrolase, Carboxylic Esterase, Chirazyme E 1, Chirazyme E-2, Chirazyme E-3, Cinnamate Esterase, Cinnamic Acid Esterase, Cinnamoyl Ester Hydrolase, Cinnamoyl Esterase, E 0231, E.C. 3.1.1.1, E.C. 3.1.1.12, EC 3.1.1.1, Egasyn, Esterase (EC 3.1.1.1), Esterase 29, Esterase 6, Esterase C4, Esterase D (E.C. 3.1.1.1), Esterase D (EC 3.1.1.1), Esterase EP10, Esterase, B-, Esterase-30, Fatty Acid Ethyl Ester Hydrolase, Fluazifop-butyl Esterase, Ketoprofen Alkyl Esterase, Ketoprofen Choline Esterase, Methyl Farnesoate Esterase, Methylbutyrase, Methylbutyrate Esterase, Monobutyrase, Naproxen Esterase, Neutral Esterase, Nonspecific Carboxylesterase, Paraben Esterase, Phthalate Ester Hydrolase, Phthalate Esterase, Pig Liver Esterase, Procaine Esterase, Propionyl Esterase, Prostaglandin A2 Methylesterase, Egasyns Proteins, SCA 50, SNSM 87, Short Chain Fatty Acid Esterase, Sterase, TAME Esterase, Texazyme PES Sp5, Thiazopyr Esterase, Triacetin Esterase, Valacyclovirase, Vitamin A Esterase, α-Carboxylesterase, α-Esterase, β-Carboxylesterase, β-Esterase, δ-Aminolevulinate Esterase
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.