missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Porcine Liver Esterase, TRC
Specifications
Specifications
| Chemical Name or Material | Porcine Liver Esterase |
| CAS | 9016-18-6 |
| Quantity | 250 mg |
| Synonym | Carboxylesterase, 1,4-Butanediol Diacrylate Esterase, 7-Amino-3-methoxy-3-cephem-4-carboxyl Ester Hydrolase, Aliesterase, Aminoacyl Esterase, B-Esterase, Biocatalytics 123, Butyrate Esterase, Butyryl Esterase, C4 Esterase, Carbonic Esterase, Carboxyesterase, Carboxyl Ester Hydrolase, Carboxyl Ester Lipase, Carboxyl Esterase, Carboxylate Esterase, Carboxylesterase 1, Carboxylesterase 2, Carboxylesterase B, Carboxylesterase ES-1, Carboxylesterases, Carboxylic Acid Esterase, Carboxylic Ester Hydrolase, Carboxylic Esterase, Chirazyme E 1, Chirazyme E-2, Chirazyme E-3, Cinnamate Esterase, Cinnamic Acid Esterase, Cinnamoyl Ester Hydrolase, Cinnamoyl Esterase, E 0231, E.C. 3.1.1.1, E.C. 3.1.1.12, EC 3.1.1.1, Egasyn, Esterase (EC 3.1.1.1), Esterase 29, Esterase 6, Esterase C4, Esterase D (E.C. 3.1.1.1), Esterase D (EC 3.1.1.1), Esterase EP10, Esterase, B-, Esterase-30, Fatty Acid Ethyl Ester Hydrolase, Fluazifop-butyl Esterase, Ketoprofen Alkyl Esterase, Ketoprofen Choline Esterase, Methyl Farnesoate Esterase, Methylbutyrase, Methylbutyrate Esterase, Monobutyrase, Naproxen Esterase, Neutral Esterase, Nonspecific Carboxylesterase, Paraben Esterase, Phthalate Ester Hydrolase, Phthalate Esterase, Pig Liver Esterase, Procaine Esterase, Propionyl Esterase, Prostaglandin A2 Methylesterase, Egasyns Proteins, SCA 50, SNSM 87, Short Chain Fatty Acid Esterase, Sterase, TAME Esterase, Texazyme PES Sp5, Thiazopyr Esterase, Triacetin Esterase, Valacyclovirase, Vitamin A Esterase, α-Carboxylesterase, α-Esterase, β-Carboxylesterase, β-Esterase, δ-Aminolevulinate Esterase |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?