missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Porcine IL-4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6543-SP
This item is not returnable.
View return policy
Description
IL-4 Monoclonal specifically detects IL-4 in Porcine samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| IL-4 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Q04745 | |
| IL4 | |
| E. coli-derived recombinant porcine IL-4 His25-Cys133 Accession # Q04745 | |
| 25 μg | |
| Primary | |
| Detects porcine IL-4 in direct ELISAs and Western blots. Does not cross-react with recombinant human IL-13 or with recombinant IL-4 from human, canine, bovine, mouse, rat, cotton rat, or feline systems. | |
| Pig | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 99605 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, IL4E12, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3565 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction