missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Porcine IL-12/IL-23 p40 Quantikine ELISA Kit
Porcine IL-12/IL-23 p40 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand: R&D Systems™ P1240
Additional Details : Weight : 0.58200kg
Description
The Porcine IL-12/IL-23 p40 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies porcine IL-12/IL-23 p40 in cell culture supernates (100 ul), serum (25 ul), edta plasma (25 ul), heparin plasma (25 ul).Specifications
46.90 - 3000pg/mL | |
HRP | |
Natural and Recombinant Porcine IL-12/IL-23 p40 | |
Cell Culture Supernates (100uL),Serum (25uL),EDTA Plasma (25uL),Heparin Plasma (25uL) | |
CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL12 p40, IL-12 subunit p40, IL12B, IL-12 B, IL-12 BNK cell stimulatory factor chain 2, interleukin 12, p40, interleukin 12 B (natural killer cell stimulatory factor 2, cytotoxic lymphocytematuration factor 2, p40), interleukin-12 beta chain, interleukin-12 subunit beta, natural killer cell stimulatory factor, 40 kD subunit, NKSF, NKSF2, NKSF2IL12, subunit p40 |
18.2pg/mL | |
Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum | |
ELISA | |
Store the unopened product at 2° to 8°C. Do not use past expiration date. |