missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Porcine IL-12/IL-23 p40 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB9121
This item is not returnable.
View return policy
Description
IL-12/IL-23 p40 Monoclonal specifically detects IL-12/IL-23 p40 in Porcine samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications
| IL-12/IL-23 p40 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Detects the p40 subunit of porcine IL-12/IL-23 in ELISAs and Western blots. In ELISAs, this antibody shows 25-35% cross-reactivity with recombinant human IL-12 and IL-23 and no cross-reactivity with recombinant mouse IL-12 or IL-23. | |
| Pig | |
| Purified |
| ELISA | |
| 116224 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| CLMF, CLMF p40, CLMF2, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL12 p40, IL-12 subunit p40, IL12B, IL-12B, IL-12BNK cell stimulatory factor chain 2, interleukin 12, p40, interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocytematuration factor 2, p40), interleukin-12 beta chain, interleukin-12 subunit beta, natural killer cell stimulatory factor, 40 kD subunit, NKSF, NKSF2, NKSF2IL12, subunit p40 | |
| Chinese hamster ovary cell line CHO-derived recombinant porcine IL-12 p70 heterodimer | |
| 500 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction