missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Porcine IL-10 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB6931-SP
This item is not returnable.
View return policy
Description
IL-10 Monoclonal specifically detects IL-10 in Porcine samples. It is validated for Neutralization.
Specifications
| IL-10 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3586 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Neutralization | |
| 148801 | |
| Neutralization 0.05-0.15 ug/mL | |
| Q29055 | |
| IL10 | |
| E. coli-derived recombinant porcine IL-10 Ser19-Asn175 Accession # Q29055 | |
| 25 μg | |
| Primary | |
| Detects porcine IL-10 in direct ELISAs. In direct ELISAs, 25-50% cross-reactivity with recombinant feline IL-10, recombinant canine IL-10, and recombinant guinea pig IL-10 is observed, approximately 10% cross-reactivity with recombinant human IL-10 is observed, less than 5% cross-reactivity with recombinant cotton rat IL-10 is observed, and no cross-reactivity with recombinant equine IL-10, recombinant mouse IL-10, recombinant rat IL-10, or recombinant viral IL-10 is observed. | |
| Pig | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction