missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Porcine GM-CSF Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB7111-100
This item is not returnable.
View return policy
Description
GM-CSF Monoclonal specifically detects GM-CSF in Porcine samples. It is validated for Neutralization.
Specifications
| GM-CSF | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, CSF2, CSF-2, GMCSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, Molgramostim, molgramostin, sargramostim | |
| E. coli-derived recombinant porcine GM-CSF Ala18-Lys144 Accession # Q29118 | |
| 100 μg | |
| Primary | |
| Detects porcine GM-CSF in direct ELISAs. | |
| Pig | |
| Purified |
| Neutralization | |
| 1202D | |
| Neutralization 0.02-0.1 ug/mL | |
| Q29118 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 1437 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction