missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Porcine Circovirus 2 Capsid Antibody [Janelia Fluor™ 669], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-26888JF669
This item is not returnable.
View return policy
Description
Porcine Circovirus 2 Capsid Polyclonal antibody specifically detects Porcine Circovirus 2 Capsid in Virus samples. It is validated for ELISA,Western Blot
Specifications
| Porcine Circovirus 2 Capsid | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Virus | |
| Purified |
| ELISA, Western Blot | |
| Janelia Fluor 669 | |
| CAP, Capsid Protein, PCV2 Capsid protein | |
| Produced in rabbits immunized with purified, recombinant Porcine Circovirus 2 Capsid (Asn42-Pro233). | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction