missing translation for 'onlineSavingsMsg'
Learn More

Porcine Circovirus 2 Capsid Antibody (GT972), Novus Biologicals™

Product Code. 18390423 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18390423 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18390423 Supplier Novus Biologicals Supplier No. NBP313604

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Porcine Circovirus 2 Capsid Monoclonal antibody specifically detects Porcine Circovirus 2 Capsid in Virus samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Porcine Circovirus 2 Capsid
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone GT972
Conjugate Unconjugated
Formulation PBS
Gene Alias CAP, Capsid Protein, PCV2 Capsid protein
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the center region of Porcine Circovirus 2 Capsid. The exact sequence is proprietary.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.