missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Porcine Circovirus 2 Capsid Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£148.00 - £301.00
Specifications
| Antigen | Porcine Circovirus 2 Capsid |
|---|---|
| Dilution | Western Blot 1:500-1:2000, ELISA 1:5000-1:10000 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043597
|
Novus Biologicals™
NBP3-26888-100ul |
100 μL |
£301.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043499
|
Novus Biologicals™
NBP3-26888-20ul |
20 μL |
£148.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Porcine Circovirus 2 Capsid Polyclonal antibody specifically detects Porcine Circovirus 2 Capsid in Virus samples. It is validated for ELISA,Western BlotSpecifications
| Porcine Circovirus 2 Capsid | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Virus | |
| CAP, Capsid Protein, PCV2 Capsid protein | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 μm filtered solution in PBS | |
| Produced in rabbits immunized with purified, recombinant Porcine Circovirus 2 Capsid (Asn42-Pro233). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title