missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Porcine CD31/PECAM-1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB3387-SP
This item is not returnable.
View return policy
Description
CD31/PECAM-1 Monoclonal specifically detects CD31/PECAM-1 in Porcine samples. It is validated for Western Blot.
Specifications
| CD31/PECAM-1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q95242 | |
| PECAM1 | |
| Mouse myeloma cell line NS0-derived recombinant porcine CD31/PECAM-1 Gln28-Lys602 (predicted) Accession # Q95242 | |
| 25 μg | |
| Primary | |
| Detects porcine CD31/PECAM-1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) CD31, rhCECAM-1, rhICAM-1, rhICAM-2, rhICAM-3, rhVCAM-1, recombinant mouse (rm) MadCAM-1, rmCD31, or rmICAM-5 is observed. | |
| Pig | |
| Purified |
| Western Blot | |
| 377521 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5175 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction