missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Haldenwanger™ Porcelain Color Test Plate
Brand: Haldenwanger™ L 61A/3
Code : 02
Additional Details : Weight : 1.63560kg
Specifications
110 x 80 x 10 mm |