Learn More
Macherey-Nagel™ Porablot Nitrocellulose membrane with supporting tissue for blotting
Description
For membrane selection and specifications, see 'Summary of porablot membrane applicability' table.
porablot NCP
The classical transfer medium for binding proteins and nucleic acids. Membrane material: 100% nitrocelluloseMembrane character: HydrophilicPore size: 0.45μmBinding capacity: 100μg/cm2Applications: Southern, Northern and Western blotting, colony and plaque transfer, dot blottingAlso available with supporting tissue, see porablot NCL.
porablot NCL
The classical transfer medium for binding proteins and nucleic acids - the supporting tissue prevents breaking, tearing, or shrinking.
- Membrane material: 100% nitrocellulose with inert supporting tissue
- Membrane character: Hydrophilic
- Pore size: 0.45μm
- Binding capacity: 100μg/cm2
- Applications: Southern, Northern and Western blotting, colony and plaque transfer, dot blottingAlso available without supporting tissue, see porablot NCP.
porablot NY amp
The amphoteric nylon membrane with supporting tissue for improved handling.
- Membrane material: High purity nylon with inert supporting tissue
- Membrane character: Amphoteric (isoelectric point at pH6.5)
- Pore size: 0.20μm
- Applications: Southern and Northern blotting, colony and plaque lifts, dot blottingAlso available with positive surface charge, see porablot NY plus.
porablot NY plus
The strongly cationic nylon membrane with supporting tissue for improved handling.
- Membrane material: High purity nylon with inert supporting tissue
- Membrane character: Cationic (in the pH range of 3 to 10)
- Pore size: 0.45μm
- Applications: Southern, Northern and Western blotting. Very well suited for electro- and vacuum blotting techniquesAlso available with positive surface charge, see porablot NY amp.
porablot PVDF
The classical transfer membrane for Western transfer, protein binding assays.
- Membrane material: Polyvinylidene difluoride (PVDF)
- Membrane character: Hydrophobic
- Pore size: 0.20μm
- Binding capacity: 50μg/cm2 to 100μg/cm2 (For large, globular proteins such as immunoglobulins, for smaller peptides the binding capacity is correspondingly larger)
- Applications: Western blotting, protein binding assays, protein sequencing
- Transfer membranes from a variety of polymeric materials with a broad range of pore sizes and surface chemistry
- Easy analysis and characterisation of biomolecules
- Conservation of expensive test solutions
- Possibility of multiple investigations with one blot
- Concentration of a sample with consequent higher detection sensitivity
- Convenient storage of results
- Outstanding band resolution due to uniform, carefully controlled pore structure
- Available in custom sizes – contact our Technical Support Team for further information Transfer membranes for DNA, RNA, bacterial colonies and protein blotting applications.
Specifications
Specifications
| Binding Capacity | 100 μg/cm2 |
| Material | Nitrocellulose |
| Pore Size | 0.45 μm |
| Dimensions (LxW) | 200 x 200 mm |
Safety and Handling
- Porablot NCL (200x200, 10 sheets)
- Danger
- Flammable solid Category 1
- H228-Flammable solid.
- MIXTURE LIST-Contain: nitrocellulose
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.