missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POR/Cytochrome P450 Reductase Antibody (OTI3F10), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73539AF350
This item is not returnable.
View return policy
Description
POR/Cytochrome P450 Reductase Monoclonal antibody specifically detects POR/Cytochrome P450 Reductase in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| POR/Cytochrome P450 Reductase | |
| Monoclonal | |
| Alexa Fluor 350 | |
| CPR, CYPORFLJ26468, DKFZp686G04235, EC 1.6.2.4, NADPH--cytochrome P450 reductase, NADPH-dependent cytochrome P450 reductase, P450 (cytochrome) oxidoreductase, P450R | |
| Full length recombinant protein of human POR/Cytochrome P450 Reductase (NP_000932) produced in HEK293T cells. | |
| 0.1 mL | |
| Breast Cancer, Stem Cell Markers | |
| 5447 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI3F10 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction