missing translation for 'onlineSavingsMsg'
Learn More

POPSO, 0.2M buffer soln., pH 8.0, Thermo Scientific™

Product Code. 15472919
Change view
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15472919 250 mL 250mL
15462919 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15472919 Supplier Thermo Scientific Alfa Aesar Supplier No. J60085.AK
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 108321-07-9 | Molecular Formula: C10H20N2Na2O8S2 | Molecular Weight: 406.376 g/mol

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108321-07-9
Molecular Formula C10H20N2Na2O8S2
Molecular Weight (g/mol) 406.376
MDL Number MFCD00070005
InChI Key XMRCLLXGMGEVHS-UHFFFAOYSA-L
PubChem CID 53393457
IUPAC Name disodium;2-hydroxy-3-[4-(2-hydroxy-3-sulfonatopropyl)piperazin-1-yl]propane-1-sulfonate
SMILES C1CN(CCN1CC(CS(=O)(=O)[O-])O)CC(CS(=O)(=O)[O-])O.[Na+].[Na+]

Specifications

Concentration 0.2 M
pH 8.0
Physical Form Liquid
Quantity 250 mL
Chemical Name or Material POPSO buffer soln.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.