Learn More
Invitrogen™ POP7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596966
Description
Immunogen sequence: MAENREPRGA VEAELDPVEY TLRKRLPSRL PRRPNDIYVN MKTDFKAQLA RCQKLLDGGA RGQNACSEIY IHGLGLAINR AINIALQLQA GSFGSLQVAA NTSTVELVDE LEPETDTREP LTRIRNNSAI HIRVFRVTPK; Positive Samples: MCF-7, SW480; Cellular Location: Cytoplasm, Cytoplasmic granule, Nucleus, nucleolus.
Component of ribonuclease P, a protein complex that generates mature tRNA molecules by cleaving their 5'-ends. Also a component of RNase MRP complex, which cleaves pre-rRNA sequences.
Specifications
| POP7 | |
| Polyclonal | |
| Unconjugated | |
| POP7 | |
| 0610037N12Rik; hPOP7; POP7; POP7 (processing of precursor, S. cerevisiae) homolog; POP7 homolog, ribonuclease P/MRP subunit; processing of precursor 7, ribonuclease P subunit; processing of precursor 7, ribonuclease P/MRP subunit; ribonuclease P protein subunit p20; Ribonucleases P/MRP protein subunit POP7 homolog; RNaseP protein p20; RPP2; RPP20 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10248 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.03 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75817 | |
| POP7 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-140 of human POP7 (NP_0058282). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.