missing translation for 'onlineSavingsMsg'
Learn More

POP7 Antibody, Novus Biologicals™

Product Code. 18384599 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18384599 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18384599 Supplier Novus Biologicals Supplier No. H00010248B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

POP7 Polyclonal antibody specifically detects POP7 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen POP7
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation PBS (pH 7.4)
Gene Accession No. AAH01430
Gene Alias 0610037N12Rik, EC 3.1.26.5, hPOP7, processing of precursor 7, ribonuclease P subunit, processing of precursor 7, ribonuclease P subunit (S. cerevisiae), processing of precursor 7, ribonuclease P/MRP subunit (S. cerevisiae), ribonuclease P protein subunit p20, Ribonucleases P/MRP protein subunit POP7 homolog, RNaseP protein p20, RPP20S. cerevisiae) homolog
Host Species Mouse
Immunogen POP7 (AAH01430, 1 a.a. - 140 a.a.) full-length human protein. MAENREPRGAVEAELDPVEYTLRKRLPSRLPRRPNDIYVNMKTDFKAQLARCQKLLDGGARGQNACSEIYIHGLGLAINRAINIALQLQAGSFGSLQVAANTSTVELVDELEPETDTREPLTRIRNNSAIHIRVFRVTPK
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10248
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.