missing translation for 'onlineSavingsMsg'
Learn More

POP7 Antibody [Alexa Fluor™ 594], Novus Biologicals™

Product Code. 30763434 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30763434 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30763434 Supplier Novus Biologicals Supplier No. NBP346934AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

POP7 Polyclonal antibody specifically detects POP7 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen POP7
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias 0610037N12Rik, EC 3.1.26.5, hPOP7, processing of precursor 7, ribonuclease P subunit, processing of precursor 7, ribonuclease P subunit (S. cerevisiae), processing of precursor 7, ribonuclease P/MRP subunit (S. cerevisiae), ribonuclease P protein subunit p20, Ribonucleases P/MRP protein subunit POP7 homolog, RNaseP protein p20, RPP20S. cerevisiae) homolog
Host Species Rabbit
Immunogen Ribonuclease P protein subunit p20. (Uniprot# O75817)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10248
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.