missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ POP5 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15898643
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15898643 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15898643 Supplier Invitrogen™ Supplier No. PA570239

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 75%.

Component of ribonuclease P, a protein complex that generates mature tRNA molecules by cleaving their 5'-ends (PubMed:11413139, PubMed:30454648). Also a component of the MRP ribonuclease complex, which cleaves pre-rRNA sequences (PubMed:28115465). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen POP5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene POP5
Gene Accession No. Q969H6
Gene Alias 1500019J17Rik; 2700077E03Rik; AD-008; AW049989; fc69d03; hPop5; HSPC004; POP5; POP5 homolog, ribonuclease P/MRP subunit; processing of precursor 5, ribonuclease P/MRP family; processing of precursor 5, ribonuclease P/MRP family (S. cerevisiae); processing of precursor 5, ribonuclease P/MRP subunit; processing of precursor 5, ribonuclease P/MRP subunit (S. cerevisiae); ribonuclease P protein 3; ribonuclease P/MRP protein subunit POP5; Ribonuclease P/MRP protein subunit POP5-like protein; Rnasep3; rpP2; RPP20; wu:fc69d03; wu:fy24c05; x0003; zgc:110616
Gene Symbols POP5
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human POP5.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51367
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.