missing translation for 'onlineSavingsMsg'
Learn More

POP4 Antibody, Novus Biologicals™

Product Code. 18383613
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18383613 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18383613 Supplier Bio-Techne Supplier No. NBP312412

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

POP4 Polyclonal antibody specifically detects POP4 in Human, Porcine, Bovine, Feline, Primate samples. It is validated for Western Blot, ELISA, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen POP4
Applications Western Blot, ELISA, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide
Gene Alias EC 3.1.26.5, processing of precursor 4, ribonuclease P/MRP subunit (S. cerevisiae), ribonuclease P protein subunit p29, RPP29hPOP4
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 170-220 on human Ribonuclease P protein subunit p29 protein.
Purification Method Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Endocrinology
Primary or Secondary Primary
Gene ID (Entrez) 10775
Target Species Human, Pig, Bovine, Feline, Primate
Content And Storage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.