missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ POP4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30562
This item is not returnable.
View return policy
Description
POP4 Polyclonal antibody specifically detects POP4 in Human samples. It is validated for Western Blot
Specifications
| POP4 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| EC 3.1.26.5, processing of precursor 4, ribonuclease P/MRP subunit (S. cerevisiae), ribonuclease P protein subunit p29, RPP29hPOP4 | |
| Between 170 and 220 | |
| 100 μg | |
| Endocrinology | |
| 10775 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction