missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POP1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £364.00
Specifications
| Antigen | POP1 |
|---|---|
| Dilution | Western Blot 1:200-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
POP1 Polyclonal antibody specifically detects POP1 in Mouse, Rat samples. It is validated for Western BlotSpecifications
| POP1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Endocrinology, Immunology | |
| PBS (pH 7.3), 50% glycerol | |
| 10940 | |
| IgG | |
| Affinity purified |
| Western Blot 1:200-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| EC 3.1.26.5, hPOP1, KIAA0061, MGC17365, processing of precursor 1, ribonuclease P/MRP subunit (S. cerevisiae), processing of precursors 1, ribonucleases P/MRP protein subunit POP1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-210 of human POP1 (NP_055844.2). MSNAKERKHAKKMRNQPTNVTLSSGFVADRGVKHHSGGEKPFQAQKQEPHPGTSRQRQTRVNPHSLPDPEVNEQSSSKGMFRKKGGWKAGPEGTSQEIPKYITASTFAQARAAEISAMLKAVTQKSSNSLVFQTLPRHMRRRAMSHNVKRLPRRLQEIAQKEAEKAVHQKKEHSKNKCHKARRCHMNRTLEFNRRQKKNIWLETHIWHAK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title