missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ POP-7™ Polymer for 3730/3730xl DNA Analyzers

POP-7™ Polymer is a separation matrix for the 3730 and 3730xl DNA Analyzers. POP-7™ Polymer is customized for DNA sequencing and fragment analysis.

Brand:  Applied Biosystems™ 4335611

6231.50 GBP valid until 2024-06-28
Use promo code "21649" to get your promotional price.



Additional Details : Weight : 0.02490kg

 View more versions of this product

Product Code. 15869776

  • £11330.00 / Each
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • A separation matrix for the 3730 and 3730xl DNA Analyzers
  • Customized for DNA sequencing and fragment analysis.
  • Pre-formulated to save time and reproducibility by ensuring consistent formulation
  • Designed to allow capillaries to be used multiple times for sequencing and fragment analysis applications
  • Not suitable for use with ABI PRISM™, 310 and 3100 series genetic analyzers as it causes syringe failure and premature array degradation
  • Sufficient for 8,000 to 12,000 samples

Use With

  • 3730/3730xl Genetic Analyzer Capillary Array, 50 cm
  • 3730 Running Buffer, 10X
  • BigDye™ Terminator v1.1 Cycle Sequencing Kit
  • BigDye™ Terminator v3.1 Cycle Sequencing Kit
  • Hi-Di™ Formamide

Order Info

Each

Not suitable for use with the 310 and 3100 series genetic analyzers. It causes syringe failure and premature array degradation.

Specifications

Specifications

98.5% up to 900 bases (for 50 cm capillary), 98.5% up to 700 bases (for 36 cm capillary)
3730 DNA Analyzer
Stability on instrument 7 days, up to 12,000 samples/28 mL
Polymer
1 hr for 36 cm capillary (std. seq.), 2 hrs for 50 cm capillary (long read seq.)
Refrigerate at 2°C to 8°C.
Standard Sequencing, Fragment Analysis (Denaturing)
POP-7™
30 x 28 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.