missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ POP-7™ Polymer for 3730/3730xl DNA Analyzers

Product Code. 15829816 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 x 28 mL
5 x 28 mL
10 x 28 mL
30 x 28 mL
Unit Size:
28mL
Each
Pack of 5
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15829816 1 x 28 mL Each
16375302 1 x 28 mL 28mL
15781816 5 x 28 mL Pack of 5
15919182 5 x 28 mL Pack of 5
15839816 10 x 28 mL Each
15869776 30 x 28 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15829816 Supplier Applied Biosystems™ Supplier No. 4363929
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Includes

Each bottle contains enough polymer for 960 samples

POP-7™ Polymer is a separation matrix for the 3730 and 3730xl DNA Analyzers. POP-7™ Polymer is customized for DNA sequencing and fragment analysis.

  • A separation matrix for the 3730 and 3730xl DNA Analyzers
  • Customized for DNA sequencing and fragment analysis.
  • Pre-formulated to save time and reproducibility by ensuring consistent formulation
  • Designed to allow capillaries to be used multiple times for sequencing and fragment analysis applications
  • Not suitable for use with ABI PRISM™, 310 and 3100 series genetic analyzers as it causes syringe failure and premature array degradation
  • Sufficient for 8,000 to 12,000 samples

Use With

  • 3730/3730xl Genetic Analyzer Capillary Array, 50 cm
  • 3730 Running Buffer, 10X
  • BigDye™ Terminator v1.1 Cycle Sequencing Kit
  • BigDye™ Terminator v3.1 Cycle Sequencing Kit
  • Hi-Di™ Formamide
TRUSTED_SUSTAINABILITY

Specifications

Accuracy 98.5% up to 900 bases (for 50 cm capillary), 98.5% up to 700 bases (for 36 cm capillary)
Content And Storage Refrigerate at 2°C to 8°C.
For Use With (Equipment) 3730xl DNA Analyzer, 3730 DNA Analyzer
For Use With (Application) Standard Sequencing, Fragment Analysis (Denaturing)
On Instrument Life Usage Stability on instrument 7 days, sufficient for 8000 to 12,000 samples
Product Line POP-7
Product Type Polymer
Quantity 1 x 28 mL
Test Time 1 hr for 36 cm capillary (std. seq.), 2 hrs for 50 cm capillary (long read seq.)
How many samples can I run with one 28 ml bottle of POP-7 polymer on my Applied Biosystems 3730 Series DNA Analyzer?

You can run 8,000-12,000 samples. These numbers may be lower if there is increased polymer consumption due to additional wizards being performed, such as the Bubble Remove and Water Wash wizards.

How long can POP-7 polymer stay on the Applied Biosystems 3730 Series DNA Analyzer?

POP-7 polymer can maintain its performance for seven days on the Applied Biosystems 3730 Series DNA Analyzer at standard lab conditions. After seven days, perform all required maintenance (including the Water Wash Wizard) and replace with fresh polymer. Do not "top off" the polymer bottle (add fresh polymer to old polymer), which can adversely affect data quality and consumable lifespan.

What polymer is supported on the Applied Biosystems 3730 Series DNA Analyzer?

Applied Biosystems supports the use of POP-7 polymer on the Applied Biosystems 3730 Series DNA Analyzer. If the instrument is running Applied Biosystems 3730 Data Collection Software v.3.1 or later, then you can also use POP-6 polymer. We do not recommend mixing and matching of various instrument polymers as each is manufactured in different processes, tested as an entire system, and guaranteed to perform to a minimum specification. Substitution of reagents may compromise the integrity of the system and cannot be supported by AB service or applications support specialists. Using unsupported polymers may adversely affect data quality, consumable lifespan, and may void existing warranties.

Does the Applied Biosystems 3130 Series use the same POP-7 Polymer as the Applied Biosystems 3730 Series?

Yes, Applied Biosystems 3130 Series Systems use the same POP-7 Polymer as Applied Biosystems 3730 series instruments, but is sold in a convenient 7 mL volume bottle which is aligned with expected Applied Biosystems 3130 Series throughput. We do not recommend customers purchase the Applied Biosystems 3730 series sized POP-7 Polymer due to the difference in throughput, limited shelf life and potential problems with pouring.

What is the white residue around the array knob on my Applied Biosystems 3730/3730xl DNA Analyzer?

When placing the array on the instrument, the last step in the process is to loosen the array knob and the instrument forces any air bubbles around the array tip through the threading and the middle of the array knob. Some polymer gets pushed through as well and when it dries up, it appears as a white, flaky residue. Use a damp, lint-free cloth to remove it. If more residue appears after the cleanup, it is possible that the array knob is not tightened enough or the array ferrule may require replacement.

There is a large bubble at the tip of the piston on my Applied Biosystems 3730/3730xl DNA Analyzer. How can I remove it?

It is not unusual to see a large bubble at the tip of the piston while performing the water wash wizard on the system. When the polymer is put back on to the system, the difference in viscosity is usually enough to dislodge the bubble and push it out. In extreme cases, it may be necessary to repeat the wash and go back and forth between the water and polymer to force it out.

When I started my run on the Applied Biosystems 3730/3730xl DNA Analyzer, an error occurred: "Leak detected" but I don't see any leaks. What else might cause this?

“Leak detected” error messages can also occur when the piston on the pump moves further than expected during the movement of the pump. The usual cause for this is a leak on the system, usually due to either an improperly tightened or improperly threaded fitting or a broken capillary. However, if this is not observed, some of the other reasons are:

1.Valve stuck in open position
2.Large air bubble under the piston
3. Polymer:
-Wrong polymer type (e.g., instrument thinks it has POP-7 on it but it really has POP-4).
-Old/degraded polymer
4. Improper/no maintenance on the system

Inspect the system for leaks. If you do not see any leaks on the system, perform a water wash using the water wash wizard and place fresh, non-expired polymer and freshly made 1X Running Buffer on the system. If the problem persists, a service call may be required.

When I started my run on the Applied Biosystems 3730/3730xl DNA Analyzer, an error occurred: "Unstable Electrophoresis current detected, check for air bubbles" but I don't see any bubbles. What else can cause this?

Some of the other causes of an Unstable Electrophoresis current detected error message are:

1.Leak on the system
2.Polymer that:
-Has expired
-Has been left on the instrument for more than the recommended time
-Is a mixture of expired polymer and non-expired polymer
3.Using Running Buffer that was:
-Diluted to 1X incorrectly
-Swapped with the water position on the
Autosampler deck
-On the instrument longer than 48 hours
-Not filled to the fill line or evaporated below the fill line
4. An arcing event that was not cleaned afterwards using the water wash wizard
5. Not performing regular maintenance on the instrument
6. Hardware issues

Inspect the system for leaks. If you do not see any leaks on the system, perform a water wash using the water wash wizard and place fresh, non-expired polymer and freshly made 1X Running Buffer on the system. If the problem persists, a service call may be required.

I have an Applied Biosystems 3730/3730xl DNA Analyzer. When do my consumables expire?

If we do not specify the expiry date or number of uses, the warranty will last for 12 months from the date of shipment.

My consumables for the Applied Biosystems 3730/3730xl DNA Analyzer have expired, but how long past expiration can I really use them?

Thermo Fisher Scientific can only provide warranty on performance on non-expired reagents. Using reagents past their expiration date is a risk and may compromise your data integrity.

I am only using the Applied Biosystems 3730/3730xl DNA Analyzer once a week. Should I leave the polymer on the instrument or put it back in the refrigerator?

Putting the polymer back at 2 - 8 degree C may extend the life of the polymer, however, we do not have any data indicating how long it might extend it. Removing the polymer and putting it back on the system can also result in a trade-off in usage. When placing it back on the system, you will need to degas the polymer for 30 - 60 min, refill the pump/block, array, and remove bubbles, which will cause you to use more polymer for maintenance than you would for running samples and may decrease the number of runs you can get out of a single polymer bottle/pouch

How long can I keep the polymer on the Applied Biosystems 3730/3730xl DNA Analyzer?

For the Applied Biosystems 3730/3730xl DNA Analyzer, the polymer can remain on the instrument for a week.


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.