missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Ponceau S, pure
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Dye Content | 70% min. |
| Infrared Spectrum | Authentic |
| Formula Weight | 760.54 |
| Grade | Pure |
| Concentration or Composition (by Analyte or Components) | Solubility •To pass testE1%1cm •≥350L g−1cm−1Lambda Max. (λ) •514nm±2nm |
| UV Spectrometry | Complies |
| Color | Bordeaux to Brown/Red |
| Quantity | 25 g |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Ponceau S |
Safety and Handling
Product Identifier
- Ponceau S
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction