missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ponceau S, MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
Specifications
Specifications
| Assay Percent Range | ∽ 75% |
| Formula Weight | 760.6 |
| Quantity | 10 g |
| Physical Form | Powder |
| Chemical Name or Material | Ponceau S |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction