missing translation for 'onlineSavingsMsg'
Learn More

Ponasterone A, TRC

Product Code. 30138599
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138599 10 mg 10mg
30137242 5 mg 5mg
30010088 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138599 Supplier Toronto Research Chemicals Supplier No. TRCP68824010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13408-56-5
Molecular Formula C27 H44 O6
Molecular Weight (g/mol) 464.63
Synonym (2β,3β,5β,22R)-2,3,14,20,22-Pentahydroxycholest-7-en-6-one, (22R)-2β,3β,14,20,22-Pentahydroxy-5β-cholest-7-en-6-one, 25-Deoxyecdysterone
IUPAC Name (2S,3R,5R,9R,10R,13R,14S,17S)-17-[(1R,2R)-1,2-dihydroxy-1,5-dimethyl-hexyl]-2,3,14-trihydroxy-10,13-dimethyl-2,3,4,5,9,11,12,15,16,17-decahydro-1H-cyclopenta[a]phenanthren-6-one
SMILES CC(C)CC[C@@H](O)[C@](C)(O)[C@H]1CC[C@@]2(O)C3=CC(=O)[C@@H]4C[C@@H](O)[C@@H](O)C[C@]4(C)[C@H]3CC[C@]12C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C27H44O6/c1-15(2)6-7-23(31)26(5,32)22-9-11-27(33)17-12-19(28)18-13-20(29)21(30)14-24(18,3)16(17)8-10-25(22,27)4/h12,15-16,18,20-23,29-33H,6-11,13-14H2,1-5H3/t16-,18-,20+,21-,22-,23+,24+,25+,26+,27+/m0/s1
Formula Weight 464.3138
Chemical Name or Material Ponasterone A
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.