missing translation for 'onlineSavingsMsg'
Learn More

PONA Standard

Product Code. 30327848
Change view
Click to view available options
Quantity:
1 mL
10 x 1 mL
Unit Size:
10mL
1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30327848 10 x 1 mL 10mL
30323819 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30327848 Supplier VHG Labs Supplier No. VHGPONA10X1
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Butene∣1-Decene∣1-Dodecene∣1-Heptene∣1-Hexene∣1-Nonene∣1-Octene∣1-Pentene∣1-Undecene∣Benzene∣Cyclohexane∣Cyclopentane∣Ethanol∣Ethyl tert-Butyl Ether (ETBE)∣Ethylbenzene∣Ethylcyclohexane∣Methanol∣Methyl tert-butyl ether∣Methylcyclohexane∣n-Butane∣n-Butylbenzene∣n-Butylcyclohexane∣n-Decane∣n-Dodecane∣n-Heptane∣n-Hexane∣n-Nonane∣n-Octane∣n-Pentane∣n-Pentylbenzene∣n-Pentylcyclohexane∣N-Propane∣n-Undecane∣Propylbenzene∣Propylcyclohexane∣tert.-Butanol∣tert-Amyl Methyl Ether (TAME)∣Toluene
CAS 106-98-9, 872-05-9, 112-41-4, 592-76-7, 592-41-6, 124-11-8
Analyte or Component Names 1-Butene∣1-Decene∣1-Dodecene∣1-Heptene∣1-Hexene∣1-Nonene∣1-Octene∣1-Pentene∣1-Undecene∣Benzene∣Cyclohexane∣Cyclopentane∣Ethanol∣Ethyl tert-Butyl Ether (ETBE)∣Ethylbenzene∣Ethylcyclohexane∣Methanol∣Methyl tert-butyl ether∣Methylcyclohexane∣n-Butane∣n-Butylbenzene∣n-Butylcyclohexane∣n-Decane∣n-Dodecane∣n-Heptane∣n-Hexane∣n-Nonane∣n-Octane∣n-Pentane∣n-Pentylbenzene∣n-Pentylcyclohexane∣N-Propane∣n-Undecane∣Propylbenzene∣Propylcyclohexane∣tert.-Butanol∣tert-Amyl Methyl Ether (TAME)∣Toluene
Shipping Condition Room Temperature
Solution Type Mixture
Solvent or Matrix Hydrocarbons
Industry Type Petrochemical Testing
Quantity 10 x 1 mL
Concentration or Composition (by Analyte or Components) N-Propane 0.5 wt%∣1-Pentene 2.5 wt%∣n-Butane 2.5 wt%∣1-Butene 2.7 wt%∣1-Decene 2.7 wt%∣1-Dodecene 2.7 wt%∣1-Heptene 2.7 wt%∣1-Hexene 2.7 wt%∣1-Nonene 2.7 wt%∣1-Octene 2.7 wt%∣1-Undecene 2.7 wt%∣Benzene 2.7 wt%∣Cyclohexane 2.7 wt%∣Ethanol 2.7 wt%∣Ethyl tert-Butyl Ether (ETBE) 2.7 wt%∣Ethylbenzene 2.7 wt%∣Ethylcyclohexane 2.7 wt%∣Methanol 2.7 wt%∣Methyl tert-butyl ether 2.7 wt%∣Methylcyclohexane 2.7 wt%∣n-Butylbenzene 2.7 wt%∣n-Butylcyclohexane 2.7 wt%∣n-Decane 2.7 wt%∣n-Dodecane 2.7 wt%∣n-Heptane 2.7 wt%∣n-Hexane 2.7 wt%∣n-Nonane 2.7 wt%∣n-Octane 2.7 wt%∣n-Pentane 2.7 wt%∣n-Pentylbenzene 2.7 wt%∣n-Pentylcyclohexane 2.7 wt%∣n-Undecane 2.7 wt%∣Propylbenzene 2.7 wt%∣Propylcyclohexane 2.7 wt%∣tert.-Butanol 2.7 wt%∣tert-Amyl Methyl Ether (TAME) 2.7 wt%∣Cyclopentane 2.8 wt%∣Toluene 2.8 wt%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.