missing translation for 'onlineSavingsMsg'
Learn More

PONA Standard

Product Code. 30323819
Change view
Click to view available options
Quantity:
1 mL
10 x 1 mL
Unit Size:
10mL
1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30323819 1 mL 1mL
30327848 10 x 1 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30323819 Supplier VHG Labs Supplier No. VHGPONA1
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Butene∣1-Decene∣1-Dodecene∣1-Heptene∣1-Hexene∣1-Nonene∣1-Octene∣1-Pentene∣1-Undecene∣Benzene∣Cyclohexane∣Cyclopentane∣Ethanol∣Ethyl tert-Butyl Ether (ETBE)∣Ethylbenzene∣Ethylcyclohexane∣Methanol∣Methyl tert-butyl ether∣Methylcyclohexane∣n-Butane∣n-Butylbenzene∣n-Butylcyclohexane∣n-Decane∣n-Dodecane∣n-Heptane∣n-Hexane∣n-Nonane∣n-Octane∣n-Pentane∣n-Pentylbenzene∣n-Pentylcyclohexane∣N-Propane∣n-Undecane∣Propylbenzene∣Propylcyclohexane∣tert.-Butanol∣tert-Amyl Methyl Ether (TAME)∣Toluene
CAS 106-98-9, 872-05-9, 112-41-4, 592-76-7, 592-41-6, 124-11-8
Analyte or Component Names 1-Butene∣1-Decene∣1-Dodecene∣1-Heptene∣1-Hexene∣1-Nonene∣1-Octene∣1-Pentene∣1-Undecene∣Benzene∣Cyclohexane∣Cyclopentane∣Ethanol∣Ethyl tert-Butyl Ether (ETBE)∣Ethylbenzene∣Ethylcyclohexane∣Methanol∣Methyl tert-butyl ether∣Methylcyclohexane∣n-Butane∣n-Butylbenzene∣n-Butylcyclohexane∣n-Decane∣n-Dodecane∣n-Heptane∣n-Hexane∣n-Nonane∣n-Octane∣n-Pentane∣n-Pentylbenzene∣n-Pentylcyclohexane∣N-Propane∣n-Undecane∣Propylbenzene∣Propylcyclohexane∣tert.-Butanol∣tert-Amyl Methyl Ether (TAME)∣Toluene
Shipping Condition Room Temperature
Solution Type Mixture
Solvent or Matrix Hydrocarbons
Industry Type Petrochemical Testing
Quantity 1 mL
Concentration or Composition (by Analyte or Components) N-Propane 0.5142 wt%∣1-Pentene 2.4744 wt%∣1-Butene 2.596 wt%∣n-Butylcyclohexane 2.6229 wt%∣n-Pentane 2.6342 wt%∣n-Octane 2.6362 wt%∣n-Pentylcyclohexane 2.6364 wt%∣tert.-Butanol 2.6377 wt%∣n-Undecane 2.6415 wt%∣Toluene 2.6434 wt%∣Ethanol 2.6459 wt%∣Cyclopentane 2.6465 wt%∣Methylcyclohexane 2.6493 wt%∣Methanol 2.6505 wt%∣n-Nonane 2.6527 wt%∣Cyclohexane 2.6578 wt%∣Benzene 2.6625 wt%∣1-Nonene 2.6628 wt%∣Methyl tert-butyl ether 2.6635 wt%∣Ethyl tert-Butyl Ether (ETBE) 2.664 wt%∣n-Decane 2.6665 wt%∣1-Dodecene 2.6673 wt%∣tert-Amyl Methyl Ether (TAME) 2.6682 wt%∣n-Hexane 2.6727 wt%∣Ethylbenzene 2.6765 wt%∣1-Undecene 2.6829 wt%∣1-Heptene 2.6849 wt%∣Ethylcyclohexane 2.6872 wt%∣1-Decene 2.6923 wt%∣1-Octene 2.6985 wt%∣1-Hexene 2.6996 wt%∣Propylcyclohexane 2.7085 wt%∣n-Heptane 2.7171 wt%∣Propylbenzene 2.7173 wt%∣n-Dodecane 2.7175 wt%∣n-Pentylbenzene 2.7293 wt%∣n-Butylbenzene 2.762 wt%∣n-Butane 2.8521 wt%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.