Learn More
Invitrogen™ PON2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595769
Description
Immunogen sequence: MGRLVAVGLL GIALALLGER LLALRNRLKA SREVESVDLP HCHLIKGIEA GSEDIDILPN GLAFFSVGLK FPGLHSFAPD KPGGILMMDL KEEKPRAREL RISRGFDLAS FNPHGISTFI DNDDTVYLFV VNHPEFKNTV EIFKFEEAEN SLLHLKTVKH ELLPSVNDIT AVGPAHFYAT NDHYFSDPFL KYLETYLNLH WANVVYYSPN; Positive Samples: HepG2, 293T; Cellular Location: Membrane, Peripheral membrane protein.
This gene encodes a member of the paraoxonase gene family, which includes three known members located adjacent to each other on the long arm of chromosome 7. The encoded protein is ubiquitously expressed in human tissues, membrane-bound, and may act as a cellular antioxidant, protecting cells from oxidative stress. Hydrolytic activity against acylhomoserine lactones, important bacterial quorum-sensing mediators, suggests the encoded protein may also play a role in defense responses to pathogenic bacteria. Mutations in this gene may be associated with vascular disease and a number of quantitative phenotypes related to diabetes. Alternatively spliced transcript variants encoding different isoforms have been described.
Specifications
| PON2 | |
| Polyclonal | |
| Unconjugated | |
| PON2 | |
| 6330405I24Rik; A esterase 2; A-esterase 2; AI481612; aromatic esterase 2; arylesterase 2; paraoxonase 2; paraoxonase nirs; PON 2; PON2; Serum aryldialkylphosphatase 2; serum paraoxonase/arylesterase 2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 5445 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.79 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q15165 | |
| PON2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-210 of human PON2 (NP_0002962). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.