missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PON2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA5142545
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize isoforms 1 and 2 (NP_000296.2; NP_001018171.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:4,000.
This gene encodes a member of the paraoxonase gene family, which includes three known members located adjacent to each other on the long arm of chromosome 7. The encoded protein is ubiquitously expressed in human tissues, membrane-bound, and may act as a cellular antioxidant, protecting cells from oxidative stress. Hydrolytic activity against acylhomoserine lactones, important bacterial quorum-sensing mediators, suggests the encoded protein may also play a role in defense responses to pathogenic bacteria. Mutations in this gene may be associated with vascular disease and a number of quantitative phenotypes related to diabetes. Alternatively spliced transcript variants encoding different isoforms have been described.Specifications
PON2 | |
Polyclonal | |
Unconjugated | |
PON2 | |
6330405I24Rik; A esterase 2; A-esterase 2; AI481612; aromatic esterase 2; arylesterase 2; paraoxonase 2; paraoxonase nirs; PON 2; PON2; Serum aryldialkylphosphatase 2; serum paraoxonase/arylesterase 2 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
5445 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG |
Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q15165 | |
PON2 | |
Peptide with sequence C-ERLLALRNRLKAS. | |
100 μg | |
Primary | |
Human | |
Liquid |