missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PON2 Antibody (SR3802), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27800-50ul
This item is not returnable.
View return policy
Description
PON2 Monoclonal antibody specifically detects PON2 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot
Specifications
| PON2 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 5445 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| SR3802 | |
| Western Blot 1:1000-1:5000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 | |
| A-esterase 2, Aromatic esterase 2, EC 3.1.1.2, EC 3.1.1.81, paraoxonase 2, paraoxonase nirs, PON 2, Serum aryldialkylphosphatase 2, serum paraoxonase/arylesterase 2 | |
| A synthesized peptide derived from human PON2 (Uniprot #: Q15165) | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction