missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PON1 Antibody (OTI2D4), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73526H
This item is not returnable.
View return policy
Description
PON1 Monoclonal specifically detects PON1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| PON1 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| PON1 | |
| Full length recombinant protein of human PON1 (NP_000437) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI2D4 | |
| Western Blot, Immunohistochemistry | |
| A-esterase 1, Aromatic esterase 1, arylesterase B-type, EC 3.1.1.2, EC 3.1.1.81, EC 3.1.8.1, ESAesterase A, K-45, MVCD5, paraoxonase 1, PON 1, PONparaoxonase B-type, serum aryldiakylphosphatase, Serum aryldialkylphosphatase 1, serum paraoxonase/arylesterase 1 | |
| Mouse | |
| Affinity Purified | |
| Alzheimers Research, Cancer, Lipid and Metabolism, Neurodegeneration, Neuroscience | |
| 5444.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction