missing translation for 'onlineSavingsMsg'
Learn More

PON1 Antibody (4G8D3), Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18412448 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18412448 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18412448 Supplier Novus Biologicals Supplier No. NBP223610AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PON1 Monoclonal antibody specifically detects PON1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ICC/IF (Negative)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PON1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, ICC/IF (Negative)
Classification Monoclonal
Clone 4G8D3
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias A-esterase 1, Aromatic esterase 1, arylesterase B-type, EC 3.1.1.2, EC 3.1.1.81, EC 3.1.8.1, ESAesterase A, K-45, MVCD5, paraoxonase 1, PON 1, PONparaoxonase B-type, serum aryldiakylphosphatase, Serum aryldialkylphosphatase 1, serum paraoxonase/arylesterase 1
Host Species Mouse
Immunogen Partial recombinant human PON1 (between residues 1-200) expressed in E. coli [Uniprot P27169]
Purification Method Ammonium sulfate precipitation
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Cancer, Lipid and Metabolism, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 5444
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.