missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POMT1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93413-0.02ml
This item is not returnable.
View return policy
Description
POMT1 Polyclonal antibody specifically detects POMT1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| POMT1 | |
| Polyclonal | |
| Western Blot 1:1000-1:2000 | |
| Dolichyl-phosphate-mannose--protein mannosyltransferase 1, EC 2.4.1, EC 2.4.1.109, FLJ37239, LGMD2K, MDDGA1, MDDGB1, MDDGC1, protein O-mannosyl-transferase 1, protein-O-mannosyltransferase 1, RT | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 310-550 of human POMT1 (NP_001070833.1). VFGKPVPCWLHSHQDTYPMIYENGRGSSHQQQVTCYPFKDVNNWWIVKDPRRHQLVVSSPPRPVRHGDMVQLVHGMTTRSLNTHDVAAPLSPHSQEVSCYIDYNISMPAQNLWRLEIVNRGSDTDVWKTILSEVRFVHVNTSAVLKLSGAHLPDWGYRQLEIVGEKLSRGYHGSTVWNVEEHRYGASQEQRERERELHSPAQVDVSRNLSFMARFSELQWRMLALRSDDSEHKYSSSPLEW | |
| 0.02 mL | |
| Cancer, Neuroscience, Signal Transduction | |
| 10585 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction