missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POMGNT2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | C3orf39 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
POMGNT2 Polyclonal specifically detects POMGNT2 in Human samples. It is validated for Western Blot.Specifications
| C3orf39 | |
| Polyclonal | |
| Rabbit | |
| NP_116195 | |
| 84892 | |
| Synthetic peptide directed towards the N terminal of human C3orf39The immunogen for this antibody is C3ORF39. Peptide sequence MHLSAVFNALLVSVLAAVLWKHVRLREHAATLEEELALSRQATEPAPALR. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| AGO61, C3orf39, chromosome 3 open reading frame 39, EC 2.4, FLJ14566, glycosyltransferase, glycosyltransferase-like domain containing 2, MDDGA8 | |
| GTDC2 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title