missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POMGNT2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35440-20ul
This item is not returnable.
View return policy
Description
POMGNT2 Polyclonal antibody specifically detects POMGNT2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| POMGNT2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| AGO61, C3orf39, chromosome 3 open reading frame 39, EC 2.4, FLJ14566, glycosyltransferase, glycosyltransferase-like domain containing 2, MDDGA8 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 160-340 of human POMGNT2 (NP_116195.2).,, Sequence:, NPDNLMHVFHDDLLPLFYTLRQFPGLAHEARLFFMEGWGEGAHFDLYKLLSPKQPLLRAQLKTLGRLLCFSHAFVGLSKITTWYQYGFVQPQGPKANILVSGNEIRQFARFMTEKLNVSHTGVPLGEEYILVFSRTQNRLILNEAELLLALAQEFQMKTVTVSLEDHTFADVVRLVSNASM | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 84892 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction