missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POMGNT1 Polyclonal Antibody

Description
Antibody detects endogenous levels of total MGAT1.
The product of the POMGNT1 gene, protein O-mannose beta-1,2-N-acetylglucosaminyltransferase, participates in O-mannosyl glycan synthesis. POMGnT1 is an N(in)/C(out) (type II) membrane protein localized in the medial-Golgi that initiates the conversion of high mannose N-glycans to complex N-glycans. Specifically, POMGnT1 is a glycosylation enzyme that participates in the synthesis of O-mannosyl glycan, a laminin-binding ligand of alpha-dystroglycan that is rarely synthesized in mammals. Mutations in the POMGNT1 gene cause muscle-eye-brain disease (MEB), an autosomal recessive disorder characterized by congenital muscular dystrophy, ocular abnormalities and lissencephaly.
Specifications
Specifications
| Antigen | POMGNT1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | POMGNT1 |
| Gene Accession No. | Q5XIN7, Q8WZA1, Q91X88 |
| Gene Alias | 0610016I07Rik; 4930467B06Rik; GnT I.2; GNTI.2; gnT-I.2; LGMD2O; MDDGA3; MDDGB3; MDDGC3; MEB; MGAT1.2; N-acetylglucosaminyltransferase I.2; O-linked mannose beta1,2-N-acetylglucosaminyltransferase; O-mannosyl N-acetylglucosaminyltransferase; OMGnT1; POMGNT1; protein O-linked mannose beta 1,2-N-acetylglucosaminyltransferase; protein O-linked mannose beta1,2-N-acetylglucosaminyltransferase; protein O-linked mannose N-acetylglucosaminyltransferase 1 (beta 1,2-); protein O-linked-mannose beta-1,2-N-acetylglucosaminyltransferase 1; protein O-linked-mannose beta-1,2-N-acetylglucosaminyltransferase 1; LOW QUALITY PROTEIN: protein O-linked-mannose beta-1,2-N-acetylglucosaminyltransferase 1; RP11-322N21.3; UDP-GlcNAc:alpha-D-mannoside beta-1,2-N-acetylglucosaminyltransferase I.2; UNQ746/PRO1475 |
| Gene Symbols | POMGNT1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?